missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Brass gauze, alloy 260, 100 mesh woven from 0.11mm (0.0045 in.) dia. wire

Brand:  Alfa Aesar™ 045208.GT

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 150 x 150mm

 View more versions of this product

Product Code. 15436825

  • £15.90 / Each


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

Brass gauze, Alloy 260
100 mesh woven from 0.11mm (0.0045 in.) dia. wire
MFCD00171255

Specifications

Brass gauze, Alloy 260
150 x 150mm
Odorless
100 Mesh
100 mesh woven from 0.11mm (0.0045 in.) dia. wire
MFCD00171255
Videos
SDS