missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Brass gauze, alloy 260, 40 mesh woven from 0.25mm (0.01in) dia wire 300x300mm

Brand:  Thermo Scientific Alfa Aesar 045165.HB

Brass gauze, alloy 260, 40 mesh woven from 0.25mm (0.01in) dia wire 300x300mm

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15456795

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.