missing translation for 'onlineSavingsMsg'
Learn More

Brass Gauze, 40 mesh woven from 0.25mm (0.01 in.) dia. wire, Alfa Aesar™

Form: Solid in various forms

Brand:  Alfa Aesar 045165.GT

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 150 x 150mm

 View more versions of this product

Product Code. 15446795

  • £11.24 / Each


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

Brass
Gauze
MFCD00171255

Specifications

Brass
150 x 150mm
Odorless
MFCD00171255
Yellow
Yellow gauze
Gauze
Videos
SDS