Learn More
Brass foil, alloy 260, 1.0mm (0.04in) thick, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045221.GT
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Brass foil, alloy has a wide range of applications, such as appliance parts, highly formable parts, ammunition components, decorative parts of furniture and cabinets, name plates, engraving plates, overlays, legend plates, lamp fixtures, fasteners, locks, hinges, flashlight shells, connectors, fittings, circuit board relays, electronic components, switches, nuts, bolts, valve stems and valve components, radiator cores and tanks, brackets, frames, fixtures, ornamental works, and used extensively in the musical instrument, electrical equipment, plumbing, central heating, seawater lines and desalination fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Brass foil, Alloy 260 | |
MFCD00171255 | |
Insoluble in water. | |
Foil |
1.0mm (0.04 in.) thick | |
150 x 150 mm | |
Odorless |
RUO – Research Use Only