missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Brass foil, alloy 260, 0.64mm (0.025 in.) thick

Brand:  Alfa Aesar™ 045203.GT

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 150 x 150mm

 View more versions of this product

Product Code. 15492197

  • £15.20 / Each


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Specifications

Chemical Identifiers

Brass foil, Alloy 260
0.64mm (0.025 in.) thick
MFCD00171255

Specifications

Brass foil, Alloy 260
0.64mm (0.025 in.) thick
Foil
150 x 150mm
Odorless
MFCD00171255
Videos
SDS