missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Brass foil, alloy 260, 0.51mm (0.02 in.) thick

Brand:  Alfa Aesar™ 045181.HB

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300 x 300mm

 View more versions of this product

Product Code. 15472187

  • £27.10 / Each


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Specifications

Chemical Identifiers

Brass foil, Alloy 260
0.51mm (0.02 in.) thick
MFCD00171255

Specifications

Brass foil, Alloy 260
0.51mm (0.02 in.) thick
Foil
300 x 300mm
Odorless
MFCD00171255
Videos
SDS