missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Brass foil, alloy 260, 0.13mm (0.005 in.) thick, 15x45cm (5.9x18in)

Brand:  Alfa Aesar™ 013504.KG

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 1 piece

 View more versions of this product

Product Code. 15436015

  • £46.00 / Each


Estimated Shipment date
from Supplier 24-01-2018
Add to basket

Specifications

Chemical Identifiers

Brass foil, Alloy 260
0.13mm (0.005 in.) thick, 15x45cm (5.9x18 in.)
MFCD00171255

Specifications

Brass foil, Alloy 260
0.13mm (0.005 in.) thick, 15x45cm (5.9x18 in.)
Foil
1 piece
Odorless
MFCD00171255
Videos
SDS