Learn More
VACUUBRAND™ Rotary Vane Vacuum Pumps: Oil Mist Filters
Oil Mist Filters and replacements for VACUUBRAND Rotary Vane Vacuum Pumps
Brand: VACUUBRAND™ 20698003
Code : 03
Additional Details : Weight : 0.81540kg
Description
- For use with VACUUBRAND Rotary Vane Vacuum Pumps
- Oil Mist Filters “FO” trap up to 99% of oil mist generated by the pump, helping preserve the laboratory environment
Specifications
Model RE2, RZ2, RE2.5, RZ2.5, RE5, RZ5, RE6, RZ6 rotary vane vacuum pump | |
Oil Mist Filter |