Learn More
VACUUBRAND™ Rotary Vane Vacuum Pumps: Inlet Separators
Inlet separators for VACUUBRAND™ Rotary Vane Vacuum Pumps.
Brand: VACUUBRAND™ 20698007
Code : A0
Additional Details : Weight : 0.80000kg
Description
- For use with VACUUBRAND Rotary Vane Vacuum Pumps
- Inlet Separators “AK” help keep condensates and particulates from entering the vacuum pump, preserving performance.
Note: For RE/RZ 2.5: For rotary vane pumps RE 2.5 (article no. 2079715x) and RZ 2.5 (article no. 2079812x) - please order Inlet Separator 20698006. For rotary vane pumps RE 2.5 (article no. 2069715x) and RZ 2.5 (article no. 2069812x) - please order Inlet Separator 20698000.
Specifications
To keep condensates and particulates from entering the vacuum pump | |
Model RE8, RE9, RE16 and RZ8, RZ9, RZ16 rotary vane vacuum pump | |
Inlet Catchpot/Separator |