Learn More
BRAND™ Transferpettor™ Replacement Seals
For use with the Transferpettor™ line, specially designed to handle viscous, foaming liquids and high vapor pressure.
Brand: BRAND™ 702866
Code : 02
Additional Details : Weight : 0.00900kg
Description
- Aids in achieving precision and accuracy
- For the most demanding pipetting operations
- Transferpettors operates on the positive displacement principle, with piston in direct contact with the aspirated liquid
- Provided with DE-M marking
- Supplied in packs of 10
Specifications
DE-M | |
200 to 1000 μL | |
10/Pk. |
HDPE | |
200 to 1000µL Transferpettor |