missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ Transferpette™ S Single Channel Fixed Volume Pipet

Uses innovative and ergonomic one-handed operation. BrandTech™ Transferpette™ S Single Channel Fixed Volume Pipet offers robust yet lightweight construction. Easy Calibration™ technology provides long lasting reliability, accuracy, and precision.

Brand:  BrandTech™ 704762

Code : 03

Additional Details : Weight : 2.26800kg

 View more versions of this product

Product Code. 10635463

  • £173.00 / Each


In Stock 5
Add to basket

Description

Description

Device is lightweight with one-handed volume adjustment.

  • Centralized pipetting button with optimized stroke length
  • Easy in-lab service
  • Robust and lightweight
  • Accepts universal tips
  • Autoclavable at 121°C/250°F (20 minutes)

Specifications

Specifications

±0.6%
ISO9001, CE, IVD, certificate of performance, individual calibration certificate
50 to 1000μL tip, 50 to 1250μL tip
3 Years
1000μL
Yes
50 to 1000μL tip, 50 to 1250μL tip
Centralized piston button design, shelf mount, 4 digit display, steam sterilizable shaft, operating manual
F1000
Videos
Documentation