Learn More
BRAND™ Transferpette™ S Adjustable Single Channel Pipet Starter Sets
Simple one-handed operation units exhibits high degree accuracy, precision, maximum versatility and reliable performance with Easy Calibration™ technology
Brand: BRAND™ 704793
Code : 03
Additional Details : Weight : 0.78400kg
Includes: Set contents:
- Three adjustable volume pipettes
- Three shelf/rack mounts
- Three filled polypropylene Tip Boxes
Description
- Available as Standard Set, Micro Set, Midsize Set and Macro Set
Specifications
Autoclavable | |
1 | |
Pipette Set, Standard | |
Demanding laboratory applications |
Laboratory | |
Transferpette | |
0.5 to 10, 10 to 100, 100 to 1000 μL | |
D-10, D-100, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes |