Learn More
BRAND™ Self-adhesive Plate Sealing Film
Securely seals multiwell plates. Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate.
Brand:  BRAND™ 781391
152.78 GBP valid until 2024-12-31
Use promo code "21898" to get your promotional price.
Code : LC
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10767564
  • £176.00 / Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
  • Easily apply to plates and remove without the use of expensive equipment.
  • Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
  • Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
    • Polyethylene
    • Polyester
    • Aluminum
    • Rayon
    • Polypropylene
    • Vinyl
Specifications
Specifications
Highly transparent
100 Sheets
Sealing Film
Polyester
ELISA, Real-time PCR