Learn More
BrandTech™ BRAND™ Self-adhesive Plate Sealing Film
Securely seals multiwell plates. BrandTech™ BRAND™ Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate. Sealing films are available in a variety of materials especially suited for storage, immunoassay, or cell and tissue culture.
Brand:  BrandTech™ 781380
UNSPSC : 41122410
Code : LB
Additional Details : Weight : 0.32000kg
 View more versions of this product
Product Code. 10085750
  • £161.00 / Pack of 100
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Description
  • Easily apply to plates and remove without the use of expensive equipment.
  • Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
  • Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
    • Polyethylene
    • Polyester
    • Aluminum
    • Rayon
    • Polypropylene
    • Vinyl
Description & Specifications
Specifications
Aluminum
Cold storage
Roll of 100 Sheets
Sealing film
Documentation
Technical Documents
Alternative Products