Learn More
BrandTech™ BRAND Transferpette™ -8/-12 Multichannel Adjustable Volume Pipette
Brand:  BrandTech™ 703630
UNSPSC : 41121503
Code : 03
Additional Details : Weight : 2.49480kg
 View more versions of this product
Product Code. 10794283
  • £1045.31 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications Description & Specifications
Specifications
<±0.6%
ISO and GLP
Yes
3 Years
20 to 200μL
12
Yes
Documentation
Technical Documents