Learn More
BrandTech™ BRAND Transferpette™ -8/-12 Multichannel Adjustable Volume Pipette
Brand:  BrandTech™ 703622
UNSPSC : 41121503
Code : 03
Additional Details : Weight : 0.77000kg
 View more versions of this product
Product Code. 10372882
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications Description & Specifications
Specifications
<±1%
ISO and GLP
3 Years
2 to 20μL
Yes
Yes
12
Documentation
Technical Documents
Alternative Products