missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ Telescoping Filling Tube for Dispensette™III, Titrette™, and seripettor™ pro

Manufactured from FEP. BrandTech™ Telescoping Filling Tube for Dispensette™III, Titrette™, seripettor™ pro adjusts easily to different size bottles without measuring or cutting. Available in a variety of lengths and fits a wide range of bottles.

Brand:  BrandTech™ 704202

Code : 03

Additional Details : Weight : 0.11340kg

 View more versions of this product

Product Code. 10420021

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Manufactured from FEP
  • Adjusts easily to different size bottles without measuring or cutting.
  • For use with Dispensette™III, Titrette™, and seripettor™ pro.
  • Available in a variety of lengths
  • Fits a wide range of bottles.
Specifications

Specifications

1Each
Telescopic Filling Tube
6mm
70 to 140mm
0.5, 1, 2, 5, 10mL
Dispensette™ III, Titrette™, seripettor™ pro
FEP
Videos
Documentation

Technical Documents