missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ Botletop Dispenser Replacement Discharge Tube

Replaces discharge tube in 5, 10mL Dispensette™ III Bottletop Dispenser, 5, 10mL Dispensette™ Organic Bottletop Dispenser, and 25, 50, 100mL Dispensette™ Organic Bottletop Dispenser. BrandTech™ Bottletop Dispenser Replacement Discharge valve includes a viewing window, hinged screw cap swings out of the way when dispensing, and is available with or without a recirculation valve.

Brand:  BrandTech™ 707936

Code : 03

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10792931

  • £44.46 / Each


Estimated Shipment
11-06-2018
Add to basket

Description

Description

  • Viewing window provides easy priming verification
  • Hinged screw cap swings out of the way when dispensing
  • Discharge valve with safety ball closes when discharge valve is not mounted
  • Available with or without recirculation valve
For use with
  • 5, 10mL Dispensette&trade III Bottletop Dispenser
  • 5, 10mL Dispensette&trade Organic Bottletop Dispenser
  • 25, 50, 100mL Dispensette&trade Organic Bottletop Dispenser
Specifications

Specifications

Yellow
5, 10mL Dispensette™ organic bottletop dispenser
90mm
Discharge Tube
Integrated valve
Standard
Videos
Documentation

Technical Documents