Learn More
BrandTech™ PLASTIBRAND™ PD-Tip™ Disposable Pipet Tips
Made of HDPE and polypropylene (polypropylene and liquid crystal polymer for 0.1mL size) without lubricants or additives that can contaminate samples. BrandTech™ PLASTIBRAND™ PD-Tip™ Disposable Pipet Tips are automatically recognized by HandyStep™ electronic and AutoRep E. Bio-Cert™ tips are independently certified free from DNA, RNase, ATP, and endotoxin for critical biological applications.
Brand:  BrandTech™ 702382
Code : 03
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 10709711
  • £55.55 / Pack of 25
In Stock 22
Add to basket
Description
Description
  • Made of high-density polyethylene and polypropylene (polypropylene and liquid crystal polymer for 0.1mL size) without lubricants or additives that can contaminate samples.
  • Tip size is automatically recognized by HandyStep™ electronic and AutoRep E.
  • Available in nonsterile and Bio-Cert™ sterile. Bio-Cert™ tips are independently certified free from DNA, RNase, ATP, and endotoxin for critical biological applications.
Specifications
Specifications
HandyStep™├żUniversal Fit Repeater Pipettor
Nonsterile
HandyStep™, Universal Fit Repeater Pipettor
50mL
HDPE/PP (Polypropylene)
Videos
Documentation