BrandTech™ PLASTIBRAND™ PD-Tip™ Disposable Pipet Tips
Made of HDPE and polypropylene (polypropylene and liquid crystal polymer for 0.1mL size) without lubricants or additives that can contaminate samples. BrandTech™ PLASTIBRAND™ PD-Tip™ Disposable Pipet Tips are automatically recognized by HandyStep™ electronic and AutoRep E. Bio-Cert™ tips are independently certified free from DNA, RNase, ATP, and endotoxin for critical biological applications.
Overview
Brand: BrandTech™
Manufacturer Part Number: 702402
UNSPSC: 41121501
Code: 03
Additional Details:
Additional Details: Weight: 0.26000kg
Product Code. 10067771
Quantity Price
1 £ 127.0 / Pack of 100
In Stock 17
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Specification
Sterility Nonsterile
Material Polypropylene and liquid crystal polymer
For Use With (Equipment) BRAND Handystep and most standard repeating pipetters
Volume (Metric) 0.1mL
  • Made of high-density polyethylene and polypropylene (polypropylene and liquid crystal polymer for 0.1mL size) without lubricants or additives that can contaminate samples.
  • Tip size is automatically recognized by HandyStep™ electronic and AutoRep E.
  • Available in nonsterile and Bio-Cert™ sterile. Bio-Cert™ tips are independently certified free from DNA, RNase, ATP, and endotoxin for critical biological applications.
Alternative Products
Documentation