missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ Flexible Discharge Tubes for Dispensette™ III Bottle-Top Dispensers

Enhances the suitability of Dispensette dispensers for various applications.

Brand:  BrandTech™ 707926

Code : 03

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10676911

  • £85.01 / Each


Estimated Shipment
22-02-2018
Add to basket

Description

Description

  • For serial dispensing such as scintillation cocktail, the flexible discharge tube preserves all important safety innovations
  • PTFE material
  • Fine tip

Order Info

Not for use with the Dispensette HF or dispensers from other manufacturers.

Specifications

Specifications

Flexible Discharge Tube
Serial dispensing such as scintillation cocktail, the flexible discharge tube preserves all important safety innovations
PTFE
Fine Tip
25 to 100mL Dispensette™ III, Dispensette™ organic bottletop dispenser
Safety handle
800mm
Videos
Documentation

Technical Documents