missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates™ pureGrade™ 384-Well Microplates
Non-treated microplates for diverse applications
Brand: BRAND™ 781620
166.87 GBP valid until 2024-12-31
Use promo code "21894" to get your promotional price.
Code : 03
Additional Details : Weight : 2.08000kg
Description
- Non-treated PS surface
- Medium binding, non-sterile
- Standard plate for most applications
- Free from endotoxins, DNase, DNA, RNase; noncytoxic
Specifications
Standard | |
16 x 24 | |
Polystyrene | |
384 | |
Non-treated | |
For homogenous assays, screenings, storage |
Without Lid | |
Clear | |
0.1 mL | |
Non-sterile | |
Flat Bottom (F-well) | |
50/Cs. |