Learn More
BrandTech™ BRANDplates™ lipoGrade™ 384-Well Microplates
Immunoassay plate optimized for the immobilization of hydrophobic molecules
Brand:  BrandTech™ 781867
Code : 03
Additional Details : Weight : 3.23000kg
 View more versions of this product
Product Code. 10216813
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • High binding for molecules with predominately hydrophobic regions such as lipoproteins or peptides
  • PS, non-sterile
  • For solid phase assays with hydrophobic molecules and liquid phase with hydrophilic molecules, respectively
  • Alternative to immunoGrade™ surface
  • Available in transparent, white or black color
  • With standard or transparent bottom
  • Modification via physical-chemical process, good shelf life at room temperature
  • Free from endotoxins, DNase, DNA, RNase, non-cytotoxic
Specifications
Specifications
384-Well Microplate
Black
Non-sterile
Flat
For immobilization of biomolecules with strongly hydrophobic regions
0.12mL
Videos