Learn More
BrandTech™ BRANDplates™ hydroGrade™ 384-Well Microplates
Immunoassay plate optimized for immobilization of hydrophilic molecules
Brand:  BrandTech™ 781801
Code : 03
Additional Details : Weight : 2.75000kg
 View more versions of this product
Product Code. 10377433
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • High affinity for molecules with hydrophilic regions such as glycoproteins, peptides and more
  • PS, nonsterile
  • For solid phase assays with hydrophilic molecules and liquid phase with hydrophobic molecules, respectively
  • Alternative to immunoGrade™ surface
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase, noncytotoxic
Specifications
Specifications
384-Well Microplate
White
Polystyrene
0.1mL
Flat
For homogeneous assays with hydrophobic molecules that remain in solution
Non-sterile
Videos