Learn More
BrandTech™ BRANDplates™ cellGrade™ premium 384-Well Microplates
For serum-free and serum-reduced cultivation of cells
Brand:  BrandTech™ 782100
Code : 03
Additional Details : Weight : 2.73000kg
 View more versions of this product
Product Code. 10583624
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • Poly-D-Lysine-equivalent surface with analogous results regarding cell growth and morphology
  • Optimized cell adhesion reduces cell damage from frequent washing
  • Alternative option to biologically coated surfaces
  • Physical/chemical treatment has long room temperature shelf life compared to biological coatings
  • Available in clear, white or black with standard or transparent F-bottom and lid
  • Sterilized by beta-radiation
  • DNA-, DNase-, RNase-free; noncytotoxic
  • Endotoxins are <0.01EU/mL
Specifications
Specifications
Premium 384-Well Standard Microplate
ANSI, SLAS
Cultivation of fastidious adherent cell lines
Polystyrene
Sterile
100μL
100μL
Flat
Clear
384 well
384
Carboxyl, Hydroxyl and free Amino groups
With Lid
Videos