Learn More
BrandTech™ BRANDplates™ cellGrade™ premium 384-Well Microplates
For serum-free and serum-reduced cultivation of cells
Brand:  BrandTech™ 782107
Code : 03
Additional Details : Weight : 3.76000kg
 View more versions of this product
Product Code. 10022743
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
Premium 384-Well Standard Microplate
ANSI, SLAS
Cultivation of fastidious adherent cell lines
Polystyrene
Sterile
120μL
120μL
Flat
Black
384 well
384
Carboxyl, Hydroxyl and free Amino groups
With Lid
Videos