Learn More
BrandTech™ BRAND™ Pipet Tips, 1 to 50uL
Fits most pipettes that use standard ultra-micro/crystal tips
Brand:  BrandTech™ 732106
Code : NEW
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15184897
  • £43.30 / Pack of 960
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Nonsterile
  • Available in either standard or ultra low retention types
  • Available either in bulk packages or in convenient rack format
  • All unopened packages are DNA-, RNase-, ATP- and endotoxin-free
  • For reverse pipetting of small volumes
Ultra low retention tips
  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface with low surface tension for minimal retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents or other difficult liquids where sample loss is possible
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention properties
Specifications
Specifications
Rack
1 to 50μL
Colorless
DNase, RNase, ATP, and Endotoxin Free
Universal Fit
Nonsterile
PP (Polypropylene)
High translucency, easy ejection and low waste, quick volume check
Videos