Learn More
BrandTech™ BRAND™ Pipet Tips, 1 to 50uL
Fits most pipettes that use standard ultra-micro/crystal tips
Brand:  BrandTech™ 732026
Code : NEW
Additional Details : Weight : 0.19000kg
 View more versions of this product
Product Code. 15124897
  • £21.75 / Pack of 1000
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Specifications
Tip
Nonsterile
Standard
Bulk
DNA, RNase, ATP and endotoxin-free
1 to 50μL
Colorless
Polypropylene
10,000 Pack
High translucency, easy ejection and low waste, quick volume check
Videos