missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrandTech™ BRANDplates lipoGrade™ 96-Well Microplates
Immunoassay plate optimized for the immobilization of hydrophobic molecules
Brand: BrandTech™ 781843
Code : 03
Additional Details : Weight : 4.16000kg
Description
- High binding for molecules with predominately hydrophobic regions such as lipoproteins or peptides
- PS, non-sterile
- Blue alphanumeric embossing on clear, white and black solid plates for easy well identification
- For solid phase assays with hydrophobic molecules and liquid phase with hydrophilic molecules, respectively
- Alternative to immunoGrade™ surface
Specifications
96-Well Assay Microplate | |
High Binding | |
ANSI, SLAS | |
Solid phase with hydrophobic molecules and liquid phase with hydrophilic molecules | |
Non-sterile | |
Round |
Yes | |
C shaped | |
Clear | |
Polystyrene | |
350μL |