missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates lipoGrade™ 96-Well Microplates
Immunoassay plate optimized for the immobilization of hydrophobic molecules
Brand: BRAND™ 781840
266.95 GBP valid until 2024-12-31
Use promo code "21894" to get your promotional price.
Code : 03
Additional Details : Weight : 4.25000kg
Description
- High binding for molecules with predominately hydrophobic regions such as lipoproteins or peptides
- PS, non-sterile
- Blue alphanumeric embossing on clear, white and black solid plates for easy well identification
- For solid phase assays with hydrophobic molecules and liquid phase with hydrophilic molecules, respectively
- Alternative to immunoGrade™ surface
Specifications
Includes Barcode | |
Standard | |
8 x 12 | |
Clear | |
96 | |
Round | |
100/Cs. |
ANSI, SLAS | |
Without Lid | |
High Binding | |
Polystyrene | |
Non-sterile | |
Solid phase with hydrophobic molecules and liquid phase with hydrophilic molecules |