Learn More
BrandTech™ BRANDplates lipoGrade™ 96-Well Microplates
Immunoassay plate optimized for the immobilization of hydrophobic molecules
Brand:  BrandTech™ 781847
Code : 03
Additional Details : Weight : 4.15000kg
 View more versions of this product
Product Code. 10698463
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • High binding for molecules with predominately hydrophobic regions such as lipoproteins or peptides
  • PS, non-sterile
  • Blue alphanumeric embossing on clear, white and black solid plates for easy well identification
  • For solid phase assays with hydrophobic molecules and liquid phase with hydrophilic molecules, respectively
  • Alternative to immunoGrade™ surface
Specifications
Specifications
96-Well Assay Microplate
High Binding
ANSI, SLAS
Solid phase with hydrophobic molecules and liquid phase with hydrophilic molecules
Non-sterile
Round
Yes
U shaped
Black
Polystyrene
330μL
Videos