Learn More
BrandTech™ BRANDplates lipoGrade™ 96-Well Microplates
Featuring a strongly hydrophobic/lipophilic surfacehomogeneous assays with hydrophilic molecules that remain in solution
Brand:  BrandTech™ 781849
Code : 03
Additional Details : Weight : 4.17000kg
 View more versions of this product
Product Code. 10174093
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • Immunoassay surface for the immobilization of biomolecules with strongly hydrophobic regions
  • Designed to minimize cross-contamination
  • Blue alphanumeric embossing for easy well identification
  • Non-sterile
Order Info
Packaging is 20 stacks of 5 plates each (100 plates). Each stack comes with one lid.
Specifications
Specifications
96-Well Microplate
ANSI, SLAS
Immobilization of hydrophobic molecules
Polystyrene
Without Lid
350μL
Non-sterile
Black
96 well
96
Round
Videos