Learn More
BrandTech™ BRANDplates lipoGrade™ 96-Well Microplates
Featuring a strongly hydrophobic/lipophilic surfacehomogeneous assays with hydrophilic molecules that remain in solution
Brand:  BrandTech™ 781850
Code : 03
Additional Details : Weight : 6.22000kg
 View more versions of this product
Product Code. 10544394
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • Immunoassay surface for the immobilization of biomolecules with strongly hydrophobic regions
  • Designed to minimize cross-contamination
  • Blue alphanumeric embossing for easy well identification
  • Non-sterile
Specifications
Specifications
96-Well Microplate
White
96 well
96
Without Lid
ANSI, SLAS
Immobilization of hydrophobic molecules
Polystyrene
Non-sterile
330μL
Videos