Learn More
BrandTech™ BRANDplates immunoGrade™ 96-Well Microplates
An excellent choice for the majority of standard ELISAs as well as solid phase immunoassays
Brand:  BrandTech™ 781726
Code : 03
Additional Details : Weight : 5.37000kg
 View more versions of this product
Product Code. 10731544
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • Optimized for the immobilization of IgG
  • High binding capacity for molecules with mixed hydrophobic and hydrophilic regions
  • Designed to minimize cross-contamination
  • Blue alphanumeric embossing for easy well identification on standard plates
  • Non-sterile
Specifications
Specifications
96-Well Microplate
White
96 well
96
With Lid
350μL
High Binding
For the majority of standard ELISAs as well as solid phase immunoassays
Polystyrene
Non-sterile
Round
Videos