BrandTech™ BRANDplates immunoGrade™ 96-Well Microplates

An excellent choice for the majority of standard ELISAs as well as solid phase immunoassays

Overview
Brand: BrandTech™

Manufacturer Part Number: 781729

UNSPSC: 41122107

Code: 03

Additional Details:
Additional Details: Weight: 4.18000kg



Product Code. 10072253

Quantity Price
1 £ 356.0 / Pack of 100
Estimated Shipment
19-06-2017
Add to basket


Description and Specification

Specification

Lid Yes
Sterile No
No. of Wells 96
For Use With (Application) For the majority of standard ELISAs as well as solid phase immunoassays
Type 96-Well Microplate
Material Polystyrene
Binding Property High
Volume (Metric) 350μL
Color Black
Well Design Round
Closure Yes
Format 96-well