Learn More
BrandTech™ BRANDplates hydroGrade™ 96-Well Microplates
Featuring a strongly hydrophilic surface for binding of hydrophilic molecules or for homogeneous assays with hydrophobic molecules that remain in solution
Brand:  BrandTech™ 781789
Code : 03
Additional Details : Weight : 4.18000kg
 View more versions of this product
Product Code. 10627523
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • Immunoassay surface with strong affinity to hydrophilic molecules, such as glycoproteins and peptides, antibodies with predominantly hydrophilic regions, and nucleic acids
  • Blue alphanumeric embossing for easy well identification
  • Non-sterile
Specifications
Specifications
96-Well Microplate
Black
96 well
96
Hydrophilic
Round
ANSI, SLAS
Immobilization of hydrophilic molecules
Polystyrene
Non-sterile
Without Lid
350μL
Videos