Learn More
BrandTech™ BRANDplates cellGrade™ premium 96-Well Microplates
Plates with surface characteristics for the most demanding cell lines
Brand:  BrandTech™ 782085
Code : 03
Additional Details : Weight : 3.18000kg
 View more versions of this product
Product Code. 10327773
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • Poly-D-Lysine-equivalent surface with analogous results regarding cell growth and morphology
  • Optimized cell adhesion reduces cell damage from frequent washing
  • Allows for serum-free and serum-reduced cultivation
  • Physical/chemical treatment has long room temperature shelf life compared to biological coatings
  • Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet requirements of Ph. Eur. and USP 29
  • DNA-, DNase-, RNase-free
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification
Specifications
Specifications
Premium 96-Well Microplate
White
96 well
96
Poly-D-Lysine Equivalent
350μL
ISO 11137, USP 29
Cultivation of cell lines with the highest demands on their environment
Polystyrene
Sterile
With Lid
Videos