Learn More
BrandTech™ BRANDplates cellGrade™ plus 96-Well Microplates
Designed for the cultivation of cells in reduced-serum media and fastidious cell lines
Brand:  BrandTech™ 782025
Code : 03
Additional Details : Weight : 3.17000kg
 View more versions of this product
Product Code. 10741734
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • Hydrophilic surface with freely-accessible amino, carboxyl and hydroxyl groups
  • Protein-like composition allows for direct cellular attachment
  • Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • DNA-, DNase-, RNase-free
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification
Specifications
Specifications
Plus 96-Well Microplate
White
96 well
96
Hydrophilic
350μL
ISO 11137, DIN EN ISO 10993
Cultivation of cells in reduced serum media and fastidious cell lines
Polystyrene
Sterile
With Lid
Videos