missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates cellGrade™ 96-Well, cellGrade plus-Treated, Flat-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:With Lid
Brand: BRAND™ 782022
139.02 GBP valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Code : 03
Additional Details : Weight : 2.70000kg
Description
- Hydrophilic surface with freely-accessible amino, carboxyl and hydroxyl groups
- Protein-like composition allows for direct cellular attachment
- Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
- Meet the requirements of Ph. Eur. and the USP 29
- DNA-, DNase-, RNase-free
- Endotoxins are <0.01 EU/mL
- Orange alphanumeric embossing for easy well identification
Order Info
Packs of 50 individually wrapped plates with lids
Specifications
Hydrophilic | |
Standard | |
8 x 12 | |
Polystyrene | |
Sterile | |
350 μL | |
Cultivation of cells in reduced serum media and fastidious cell lines | |
1/Pk. |
ISO 11137, DIN EN ISO 10993 | |
With Lid | |
Clear | |
96 | |
cellGrade Plus | |
Flat Bottom (F-well) | |
50/Cs. | |
Clear |