Learn More
BrandTech™ BRANDplates cellGrade™ plus 96-Well Microplates
Designed for the cultivation of cells in reduced-serum media and fastidious cell lines
Brand:  BrandTech™ 782030
Code : 03
Additional Details : Weight : 3.62000kg
 View more versions of this product
Product Code. 10564584
  • £745.00 / Pack of 50
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Hydrophilic surface with freely-accessible amino, carboxyl and hydroxyl groups
  • Protein-like composition allows for direct cellular attachment
  • Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • DNA-, DNase-, RNase-free
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification
Specifications
Specifications
Plus 96-Well Microplate
White
96 well
96
Hydrophilic
330μL
ISO 11137, DIN EN ISO 10993
Cultivation of cells in reduced serum media and fastidious cell lines
Polystyrene
Sterile
With Lid
Videos