Learn More
BRAND™ Self-adhesive Plate Sealing Film
Securely seals multiwell plates. Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate.
Brand: BRAND™ 701365
197.19 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Code : 03
Additional Details : Weight : 0.11700kg
Description
- Easily apply to plates and remove without the use of expensive equipment.
- Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
- Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
- Polyethylene
- Polyester
- Aluminum
- Rayon
- Polypropylene
- Vinyl
Specifications
Gas permeable | |
Sterile | |
Cell and tissue culture |
Rayon | |
50 Sheets | |
Sealing Film |