Learn More
BrandTech™ BRAND™ BIO-CERT™ Pipet Tips, 1 to 50uL
DNA, RNase-, ATP- and endotoxin-free
Brand:  BrandTech™ 732226
Code : NEW
Additional Details : Weight : 1.08000kg
 View more versions of this product
Product Code. 15154917
  • £82.40 / Pack of 960
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • High purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile tips are free from biological contaminants
  • Available in either standard or ultra low retention types
  • Available in convenient, ecological rack format
Ultra low retention tips
  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can increase sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention properties
Specifications
Specifications
Rack
1 to 50μL
PP (Polypropylene)
Universal Fit
Sterile
DNase, RNase, ATP, and Endotoxin Free
Videos