missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ BRAND™ BIO-CERT™ Filter Tips, 5 to 100uL

Manufactured from high-purity, oleamide- and diHEMDA-free polypropylene. BrandTech™ BRAND™ BIO-CERT™ Filter tips, 5 to 100μL allow sample recovery with wetted filters. Sterile tips are free of biological contaminants. Thin-walled for high translucency, easy ejection, and low waste. Graduations marked at 50 and 100μL for quick volume assessment. Available in either standard or ultra low retention types.

Brand:  BrandTech™ 732830

Code : NEW

Additional Details : Weight : 1.25000kg

 View more versions of this product

Product Code. 15114997

  • £110.00 / Pack of 960


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Non-sealing PE filters allow sample recovery with wetted filters
  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile tips are DNA-, RNase, ATP- and endotoxin-free
  • Thin-walled for high translucency, easy ejection and low waste
  • Graduations at 50 and 100μL for quick volume check
  • Available in either standard or ultra low retention types
  • Available in convenient, ecological rack format, racked tips are colorless and placed into green-colored tip tray

Ultra low retention tips

  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain protein, nucleic acid, detergents or other difficult liquids that can result in sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention and filter properties

Specifications

Specifications

Filter Tip
Sterile
50, 100μL
Ultra Low Retention
Rack, TipBox 10 x 96
5.3cm
For pipettes with yellow color code
5 to 100μL
Colorless
Yes
Polypropylene
960 Pack
DNA, RNase, ATP and endotoxin-free
Videos