missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ BRAND™ BIO-CERT™ Filter Tips, 2 to 20uL

Used for pipettes yellow color code

Brand:  BrandTech™ 732628

Code : NEW

Additional Details : Weight : 0.90000kg

 View more versions of this product

Product Code. 15104967

  • £86.00 / Pack of 960


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile and free of biological contaminants
  • Non-self sealing filters allow sample recovery if filter is wetted
  • Thin-walled
  • With PE filter to protect both sample and pipette
  • Available in either standard or ultra low retention types
  • Available in convenient rack format, racked tips are colorless and placed into blue-colored tip tray

Ultra low retention tips

  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can increase sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention or filter properties

Specifications

Specifications

Rack
Rack
2 to 20μL
Sterile
Yellow
PP (Polypropylene)
960 Pack
Universal Fit
Filtered
Colorless
20μL
5cm
Videos