missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ BRAND™ Pipet Tips, 1 to 10mL

Precision-molded from high purity, DiHEMDA- and oleamide-free, virgin polypropylene. BrandTech™ BRAND™ Pipet Tips, 1 to 10mL are ideal for working with the microliter pipette Transferpette™ S 10mL. Compatible with Eppendorf™ systems.

Brand:  BrandTech™ 702603

Code : NEW

Additional Details : Weight : 0.69000kg

 View more versions of this product

Product Code. 15124887

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Manufactured in an environmentally friendly manner using only high-purity polypropylene, free from oleamide and diHEMDA.
  • Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.
  • Ideal for working with the microliter pipet Transferpette™ S 10mL.
    • Compatible with Eppendorf™ systems.
Specifications

Specifications

Tip
Nonsterile
Polypropylene
200 Pack
DiHEMDA- and oleamide-free
1 to 10μL
Standard
Bulk, 2 bags of 100 each
15.65cm
Videos