missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bradykinin Acetate Salt

For cellular and molecular biology applications

Brand:  Thermo Scientific Chemicals J60345.MB

Code : NEW

Additional Details : CAS Number : 6846-03-3 Weight : 0.00025kg

 View more versions of this product

Product Code. 15463129

  • £187.00 / 25mg
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Bradykinin Acetate elicits vasoconstriction, a systemic blood pressure reflex, a stimulation of respiration, and contraction of the nictitating membrane. Bradykinin plays an important role in the regulation of fluid and electrolyte balance, smooth muscle contraction, vasodilation and capillary permeability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

HPLC
Powder
Bradykinin acetate salt
RUO - research use only
-30°C to -10°C
1120.21
25 mg
Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg-OH
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only