missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bradykinin (2-7)

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J66151.MB

Code : NEW

Additional Details : Weight : 0.00025kg

 View more versions of this product

Product Code. 15467399

  • £90.42 / 25mg


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Disclaimers:

For Research Use Only


Specifications

Specifications

600.66
25mg
Pro-Pro-Gly-Phe-Ser-Pro
-30° to -10°C
HPLC
Solid
RUO
Bradykinin (2-7)
Videos
SDS