missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ToughStripe™ Floor Arrows

ToughStripe™ Floor Arrows are extremely durable die-cut floor marking shapes made of rugged polyester with ultra-aggressive adhesive that holds up against forklift traffic.

Brand:  Brady™ 104419

Code : NEW

Additional Details : Weight : 1.18620kg

 View more versions of this product

Product Code. 11767084

  • £193.00 / 1 pack


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Visible floor-marking improves safety and organization in almost any industrial environment
  • High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Can be easily applied by one person working alone
Specifications

Specifications

ANSI A1264.2-2006, OSHA 1910.22
High-gloss
100 Pack
Permanent Rubber
Ideal for marking aisles, passageways and storage locations and pointing out important safety equipment and egress paths
54°C
-18°C
ToughStripe™ floor marking tape
-18 °C-54 °C
Green
Polyester
Arrow
ToughStripe Diecut Shapes (L's, T's, +'s, footprints and more)
203.20 mm
Arrow
2mil
76.2 mm
76.20 mm203.20 mm
Videos