Learn More
BPOZ Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557119
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MLDEDTVVGV WRVAKLFRLA RLEDQCTEYM AKVIEKLVER EDFVEAVKEE AAAVAARQET DSIPLVDDIR FHVASTVQTY SAIEEAQQRL RALEDLLVSI GL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.
BPOZ Polyclonal antibody specifically detects BPOZ in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
BPOZ | |
Polyclonal | |
Unconjugated | |
ABTB1 | |
ABTB1; abtb1 {ECO:0000312; AI847549; ankyrin repeat and BTB (POZ) domain containing 1; ankyrin repeat and BTB domain containing 1; ankyrin repeat and BTB/POZ domain-containing protein 1; BC003234; BPOZ; BTB3; BTBD21; EF1ABP; elongation factor 1A-binding protein; EMBL:AAH83579.1}; mFLJ00331 protein; MGC20585; PP2259 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80325 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969K4 | |
ABTB1 | |
Recombinant protein corresponding to Human BPOZ | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only