missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BPIL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558048
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NVLSRIAEIY ILSQPFMVRI MATEPPIINL QPGNFTLDIP ASIMMLTQPK NSTVETIVSM DFVASTSVGL VILGQRLVCS LSLNRFR Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 75%.
BPIL2 Polyclonal antibody specifically detects BPIL2 in Human samples. It is validated for Immunohistochemistry.Specifications
BPIL2 | |
Polyclonal | |
Unconjugated | |
BPIFC | |
4732454E24; bactericidal/permeability-increasing protein-like 2; BPI fold containing family C; BPI fold-containing family C protein; Bpifc; Bpil2; BPI-like 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
254240 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NFQ6 | |
BPIFC | |
Recombinant protein corresponding to Human BPIL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only